Learn More
Biomatik Corporation Human Complement C4-B (C4B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03398
Encompass_Preferred
The Human Complement C4-B (C4B) ELISA Kit allows for quantitative determination of Complement C4-B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 4.7-300 ng/mL. Sensitivity: 2.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P0C0L5. Cross Activity: No significant cross-reactivity or interference between Complement C4-B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2204