Learn More
Biomatik Corporation Mouse Complement C4-B (C4B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03399
Encompass_Preferred
Mouse Complement C4-B (C4B) ELISA Kit for detecting C4B in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.72ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01029. Gene ID: 12268. Target Synonyms: CO4; CPAMD3; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; Complement component 4B, Chido blood group. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2205