Learn More
Biomatik Corporation Human Complement Component 5 (C5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03401
Encompass_Preferred
The Human Complement Component 5 (C5) ELISA Kit allows for quantitative determination of Complement Component 5 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.45 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01031. Cross Activity: No significant cross-reactivity or interference between Complement Component 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2207