Learn More
Biomatik Corporation Mouse Complement Component 5 (C5) ELISA Kit, 96 tests
Catalog No. 501502208
Encompass_Preferred
Mouse Complement Component 5 (C5) ELISA Kit for detecting C5 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800ng/ml. Sensitivity: 4.4ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06684. Gene ID: 15139. Target Synonyms: CPAMD4; C5a anaphylatoxin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2208 Supplier Biomatik Corporation Supplier No. EKU03402
Mouse Complement Component 5 (C5) ELISA Kit for detecting C5 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800ng/ml. Sensitivity: 4.4ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06684. Gene ID: 15139. Target Synonyms: CPAMD4; C5a anaphylatoxin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.