Learn More
Biomatik Corporation Rat Complement Component 5 (C5) ELISA Kit, 96 tests
Catalog No. 501502209
Encompass_Preferred
Rat Complement Component 5 (C5) ELISA Kit for detecting C5 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.142ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A096P6L9. Target Synonyms: CPAMD4; C5a anaphylatoxin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2209 Supplier Biomatik Corporation Supplier No. EKU03403
Rat Complement Component 5 (C5) ELISA Kit for detecting C5 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.142ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A096P6L9. Target Synonyms: CPAMD4; C5a anaphylatoxin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.