Learn More
Biomatik Corporation Dog Complement Component 5a (C5a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03405
Encompass_Preferred
The Dog Complement Component 5a (C5a) ELISA Kit allows for quantitative determination of Complement Component 5a in Serum and Plasma. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.88 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9TSW9. Cross Activity: No significant cross-reactivity or interference between Complement Component 5a and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2210