Learn More
Biomatik Corporation Rat Complement Component 5a (C5a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03409
Encompass_Preferred
Rat Complement Component 5a (C5a) ELISA Kit for detecting C5a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08650. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2214