Learn More
Biomatik Corporation Mouse Complement Component 5a Receptor 1 (C5aR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03411
Encompass_Preferred
Mouse Complement Component 5a Receptor 1 (C5aR1) ELISA Kit for detecting C5aR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30993. Gene ID: 12273. Target Synonyms: CD88; C5A; C5-AR1; C5R1; C5a anaphylatoxin chemotactic receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2215