Learn More
Biomatik Corporation Human Complement Factor B (CFB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03417
Encompass_Preferred
The Human Complement Factor B (CFB) ELISA Kit allows for quantitative determination of Complement Factor B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.25-80 ng/mL. Sensitivity: 0.61 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00751. Cross Activity: No significant cross-reactivity or interference between Complement Factor B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2218