Learn More
Biomatik Corporation Human Complement Factor H Related Protein 5 (CFHR5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03428
Encompass_Preferred
The Human Complement Factor H Related Protein 5 (CFHR5) ELISA Kit allows for quantitative determination of Complement Factor H Related Protein 5 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 14.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BXR6. Cross Activity: No significant cross-reactivity or interference between Complement Factor H Related Protein 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2226