Learn More
Biomatik Corporation Human Complement Factor P (CFP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03430
Encompass_Preferred
The Human Complement Factor P (CFP) ELISA Kit allows for quantitative determination of Complement Factor P in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 ng/mL. Sensitivity: 6.5 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P27918. Cross Activity: No significant cross-reactivity or interference between Complement Factor P and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2228