Learn More
Biomatik Corporation Human Complement Receptor 1, Erythrocyte (CR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03431
Encompass_Preferred
The Human Complement Receptor 1, Erythrocyte (CR1) ELISA Kit allows for quantitative determination of Complement Receptor 1, Erythrocyte in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17927. Cross Activity: No significant cross-reactivity or interference between Complement Receptor 1, Erythrocyte and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2229