Learn More
Biomatik Corporation Human Complexin 2 (CPLX2) ELISA Kit, 96 tests
Catalog No. 501502230
Encompass_Preferred
The Human Complexin 2 (CPLX2) ELISA Kit allows for quantitative determination of Complexin 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 246.9-20000 pg/mL. Sensitivity: 86.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q6PUV4. Cross Activity: No significant cross-reactivity or interference between Complexin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2230 Supplier Biomatik Corporation Supplier No. EKU03434
The Human Complexin 2 (CPLX2) ELISA Kit allows for quantitative determination of Complexin 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 246.9-20000 pg/mL. Sensitivity: 86.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q6PUV4. Cross Activity: No significant cross-reactivity or interference between Complexin 2 and analogues was observed. Restriction: For Research Use Only.