Learn More
Biomatik Corporation Human Connective Tissue Growth Factor (CTGF) ELISA Kit, 96 tests
Catalog No. 501502231
Encompass_Preferred
The Human Connective Tissue Growth Factor (CTGF) ELISA Kit allows for quantitative determination of Connective Tissue Growth Factor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29279. Cross Activity: No significant cross-reactivity or interference between Connective Tissue Growth Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2231 Supplier Biomatik Corporation Supplier No. EKU03436
The Human Connective Tissue Growth Factor (CTGF) ELISA Kit allows for quantitative determination of Connective Tissue Growth Factor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29279. Cross Activity: No significant cross-reactivity or interference between Connective Tissue Growth Factor and analogues was observed. Restriction: For Research Use Only.