Learn More
Biomatik Corporation Human Contactin 4 (CNTN4) ELISA Kit, 96 tests
Catalog No. 501502238
Encompass_Preferred
The Human Contactin 4 (CNTN4) ELISA Kit allows for quantitative determination of Contactin 4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IWV2. Cross Activity: No significant cross-reactivity or interference between Contactin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2238 Supplier Biomatik Corporation Supplier No. EKU03449
The Human Contactin 4 (CNTN4) ELISA Kit allows for quantitative determination of Contactin 4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IWV2. Cross Activity: No significant cross-reactivity or interference between Contactin 4 and analogues was observed. Restriction: For Research Use Only.