Learn More
Biomatik Corporation Mouse Copeptin (CPP) ELISA Kit, 96 tests
Catalog No. 501502241
Encompass_Preferred
Mouse Copeptin (CPP) ELISA Kit for detecting CPP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.16pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P35455. Gene ID: 11998. Target Synonyms: C-Terminal Provasopressin; Vasopressin-Neurophysin 2-Copeptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2241 Supplier Biomatik Corporation Supplier No. EKU03454
Mouse Copeptin (CPP) ELISA Kit for detecting CPP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.16pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P35455. Gene ID: 11998. Target Synonyms: C-Terminal Provasopressin; Vasopressin-Neurophysin 2-Copeptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.