Learn More
Biomatik Corporation Human Coronin 1A (CORO1A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03460
Encompass_Preferred
The Human Coronin 1A (CORO1A) ELISA Kit allows for quantitative determination of Coronin 1A in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.119 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P31146. Cross Activity: No significant cross-reactivity or interference between Coronin 1A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2243