Learn More
Biomatik Corporation Mouse Corticosteroid Binding Globulin (CBG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03463
Encompass_Preferred
Mouse Corticosteroid Binding Globulin (CBG) ELISA Kit for detecting CBG in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06770. Gene ID: 12401. Target Synonyms: SERPINA6; Transcortin; Serpin A6; Serpin Peptidase Inhibitor,Clade A,Member 6 (Alpha-1 Antiproteinase; Antitrypsin). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2245