Learn More
Biomatik Corporation Corticosterone (Cort) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03465
Encompass_Preferred
The Corticosterone (Cort) ELISA Kit allows for quantitative determination of Corticosterone in Serum, Plasma, Urine and other biological fluids. Species Reactivity: All species. Detection Range: 1.23-100 ng/mL. Sensitivity: 0.51 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Corticosterone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2247