Learn More
Biomatik Corporation Human Corticotropin Releasing Hormone (CRH) ELISA Kit, 96 tests
Catalog No. 501502248
Encompass_Preferred
The Human Corticotropin Releasing Hormone (CRH) ELISA Kit allows for quantitative determination of Corticotropin Releasing Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.91 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P06850. Cross Activity: No significant cross-reactivity or interference between Corticotropin Releasing Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2248 Supplier Biomatik Corporation Supplier No. EKU03467
The Human Corticotropin Releasing Hormone (CRH) ELISA Kit allows for quantitative determination of Corticotropin Releasing Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.91 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P06850. Cross Activity: No significant cross-reactivity or interference between Corticotropin Releasing Hormone and analogues was observed. Restriction: For Research Use Only.