Learn More
Biomatik Corporation Mouse Corticotropin Releasing Hormone (CRH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03468
Encompass_Preferred
Mouse Corticotropin Releasing Factor (CRF) ELISA Kit for detecting CRF in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.05pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q8CIT0. Gene ID: 12918. Target Synonyms: CRH; Corticotropin Releasing Hormone; Corticoliberin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2249