Learn More
Biomatik Corporation Cortisol (Cor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03476
Encompass_Preferred
The Cortisol (Cor) ELISA Kit allows for quantitative determination of Cortisol in Serum, Plasma, Urine and other biological fluids. Species Reactivity: All species. Detection Range: 12.35-1000 ng/mL. Sensitivity: 5.15 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Cortisol and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2251