Learn More
Biomatik Corporation Mouse Creatine Kinase, Brain (CKB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03489
Encompass_Preferred
Mouse Creatine Kinase B (CK-BB) ELISA Kit for detecting CK-BB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04447. Gene ID: 12709. Target Synonyms: CKB; CKBB; CK-B; CK-1; Creatine Kinase, Brain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2255