Learn More
Biomatik Corporation Human Creatine Kinase, Muscle (CKM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03492
Encompass_Preferred
The Human Creatine Kinase, Muscle (CKM) ELISA Kit allows for quantitative determination of Creatine Kinase, Muscle in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.61 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P06732. Cross Activity: No significant cross-reactivity or interference between Creatine Kinase, Muscle and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2258