Learn More
Biomatik Corporation Mouse Creatine Kinase, Muscle (CKM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03493
Encompass_Preferred
Mouse Creatine Kinase, Muscle (CKM) ELISA Kit for detecting CKM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07310. Gene ID: 12715. Target Synonyms: CK-MM; CKMM; CK3; CK-M; MM-CK; M-CK; Creatine kinase M-type, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2259