Learn More
Biomatik Corporation Human CREB Regulated Transcription Coactivator 3 (CRTC3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03498
Encompass_Preferred
The Human CREB Regulated Transcription Coactivator 3 (CRTC3) ELISA Kit allows for quantitative determination of CREB Regulated Transcription Coactivator 3 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6UUV7. Cross Activity: No significant cross-reactivity or interference between CREB Regulated Transcription Coactivator 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2261