Learn More
Biomatik Corporation Rat Cross Linked C-Telopeptide of Type I Collagen (CTXI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03504
Encompass_Preferred
Rat Cross Linked C-Telopeptide Of Type I Collagen (CTXI) ELISA Kit for detecting CTXI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 50.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02454; P02466. Gene ID: 29393; 84352. Target Synonyms: CTX-I; CTX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2264