Learn More
Biomatik Corporation Rat Cross Linked C-Telopeptide of Type II Collagen (CTXII) ELISA Kit, 96 tests
Catalog No. 501502266
Encompass_Preferred
Rat Cross Linked C-Telopeptide Of Type II Collagen (CTXII) ELISA Kit for detecting CTXII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 98.8-8,000pg/ml. Sensitivity: 35.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P05539. Gene ID: 25412. Target Synonyms: CTX-II; CTX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2266 Supplier Biomatik Corporation Supplier No. EKU03507
Rat Cross Linked C-Telopeptide Of Type II Collagen (CTXII) ELISA Kit for detecting CTXII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 98.8-8,000pg/ml. Sensitivity: 35.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P05539. Gene ID: 25412. Target Synonyms: CTX-II; CTX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.