Learn More
Biomatik Corporation Human Cross Linked N-Telopeptide of Type I Collagen (NTXI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03509
Encompass_Preferred
The Human Cross Linked N-Telopeptide of Type I Collagen (NTXI) ELISA Kit allows for quantitative determination of Cross Linked N-Telopeptide of Type I Collagen in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.47-200 ng/mL. Sensitivity: 0.94 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Cross Linked N-Telopeptide of Type I Collagen and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2267