Learn More
Biomatik Corporation Human C-Type Lectin Domain Family 2, Member C (CLEC2C) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03517
Encompass_Preferred
The Human C-Type Lectin Domain Family 2, Member C (CLEC2C) ELISA Kit allows for quantitative determination of C-Type Lectin Domain Family 2, Member C in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q07108. Cross Activity: No significant cross-reactivity or interference between C-Type Lectin Domain Family 2, Member C and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2269