Learn More
Biomatik Corporation Rat C-Type Natriuretic Peptide (CNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03521
Encompass_Preferred
Rat C-Type Natriuretic Peptide (CNP) ELISA Kit for detecting CNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55207. Gene ID: 114593. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2272