Learn More
Biomatik Corporation Human Cyclophilin B (CYPB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03552
Encompass_Preferred
The Human Cyclophilin B (CYPB) ELISA Kit allows for quantitative determination of Cyclophilin B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 312-5000 pg/mL. Sensitivity: 122 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23284. Cross Activity: No significant cross-reactivity or interference between Cyclophilin B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2280