Learn More
Biomatik Corporation Mouse Cyclophilin B (CYPB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03553
Encompass_Preferred
Mouse Cyclophilin B (CYPB) ELISA Kit for detecting CYPB in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24369. Gene ID: 19035. Target Synonyms: PPIB; PPI-B; CYP-S1; CyP-B; SCYLP; PPIase B; Rotamase B; S-cyclophilin; Peptidylprolyl Isomerase B; Peptidyl-Prolyl Cis-Trans Isomerase B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2281