Learn More
Biomatik Corporation Human Cystatin 1 (CST1) ELISA Kit, 96 tests
Catalog No. 501502283
Encompass_Preferred
The Human Cystatin 1 (CST1) ELISA Kit allows for quantitative determination of Cystatin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01037. Cross Activity: No significant cross-reactivity or interference between Cystatin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2283 Supplier Biomatik Corporation Supplier No. EKU03559
The Human Cystatin 1 (CST1) ELISA Kit allows for quantitative determination of Cystatin 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01037. Cross Activity: No significant cross-reactivity or interference between Cystatin 1 and analogues was observed. Restriction: For Research Use Only.