Learn More
Biomatik Corporation Mouse Cystatin 3 (CST3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03562
Encompass_Preferred
Mouse Cystatin C (Cys-C) ELISA Kit for detecting Cys-C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.132ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21460. Gene ID: 13010. Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2286