Learn More
Biomatik Corporation Human Cystatin 4 (CST4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03565
Encompass_Preferred
The Human Cystatin 4 (CST4) ELISA Kit allows for quantitative determination of Cystatin 4 in Serum, Plasma, Saliva, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.066 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01036. Cross Activity: No significant cross-reactivity or interference between Cystatin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2287