Learn More
Biomatik Corporation Human Cysteine Rich Transmembrane BMP Regulator 1 (CRIM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03577
Encompass_Preferred
The Human Cysteine Rich Transmembrane BMP Regulator 1 (CRIM1) ELISA Kit allows for quantitative determination of Cysteine Rich Transmembrane BMP Regulator 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NZV1. Cross Activity: No significant cross-reactivity or interference between Cysteine Rich Transmembrane BMP Regulator 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2293