Learn More
Biomatik Corporation Human Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03581
Encompass_Preferred
The Human Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) ELISA Kit allows for quantitative determination of Cystic Fibrosis Transmembrane Conductance Regulator in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13569. Cross Activity: No significant cross-reactivity or interference between Cystic Fibrosis Transmembrane Conductance Regulator and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2294