Learn More
Biomatik Corporation Human Cytochrome b-245 Beta Polypeptide (CYBb) ELISA Kit, 96 tests
Catalog No. 501502295
Encompass_Preferred
The Human Cytochrome b-245 Beta Polypeptide (CYBb) ELISA Kit allows for quantitative determination of Cytochrome b-245 Beta Polypeptide in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04839. Cross Activity: No significant cross-reactivity or interference between Cytochrome b-245 Beta Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2295 Supplier Biomatik Corporation Supplier No. EKU03583
The Human Cytochrome b-245 Beta Polypeptide (CYBb) ELISA Kit allows for quantitative determination of Cytochrome b-245 Beta Polypeptide in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04839. Cross Activity: No significant cross-reactivity or interference between Cytochrome b-245 Beta Polypeptide and analogues was observed. Restriction: For Research Use Only.