Learn More
Biomatik Corporation Human Cytochrome P450 1A1 (CYP1A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03590
Encompass_Preferred
The Human Cytochrome P450 1A1 (CYP1A1) ELISA Kit allows for quantitative determination of Cytochrome P450 1A1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04798. Cross Activity: No significant cross-reactivity or interference between Cytochrome P450 1A1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2296