Learn More
Biomatik Corporation Human Cytochrome P450 2E1 (CYP2E1) ELISA Kit, 96 tests
Catalog No. 501502298
Encompass_Preferred
The Human Cytochrome P450 2E1 (CYP2E1) ELISA Kit allows for quantitative determination of Cytochrome P450 2E1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05181. Cross Activity: No significant cross-reactivity or interference between Cytochrome P450 2E1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2298 Supplier Biomatik Corporation Supplier No. EKU03604
The Human Cytochrome P450 2E1 (CYP2E1) ELISA Kit allows for quantitative determination of Cytochrome P450 2E1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05181. Cross Activity: No significant cross-reactivity or interference between Cytochrome P450 2E1 and analogues was observed. Restriction: For Research Use Only.