Learn More
Biomatik Corporation Mouse Cytochrome P450 2E1 (CYP2E1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03605
Encompass_Preferred
Mouse Cytochrome P450 2E1 (CYP2E1) ELISA Kit for detecting CYP2E1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05421. Gene ID: 13106. Target Synonyms: CPE1; CYPIIE1; P450-J; P450C2E; 4-nitrophenol 2-hydroxylase; Cytochrome P450 Family 2 Subfamily E 1; Cytochrome P450,Subfamily IIE (Ethanol-Inducible) Polypeptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2299