Learn More
Biomatik Corporation Human Cytochrome P450 3A4 (CYP3A4) ELISA Kit, 96 tests
Catalog No. 501502300
Encompass_Preferred
The Human Cytochrome P450 3A4 (CYP3A4) ELISA Kit allows for quantitative determination of Cytochrome P450 3A4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08684. Cross Activity: No significant cross-reactivity or interference between Cytochrome P450 3A4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2300 Supplier Biomatik Corporation Supplier No. EKU03607
The Human Cytochrome P450 3A4 (CYP3A4) ELISA Kit allows for quantitative determination of Cytochrome P450 3A4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08684. Cross Activity: No significant cross-reactivity or interference between Cytochrome P450 3A4 and analogues was observed. Restriction: For Research Use Only.