Learn More
Biomatik Corporation Rat Cytochrome P450 7A1 (CYP7A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03611
Encompass_Preferred
Rat Cytochrome P450 7A1 (CYP7A1) ELISA Kit for detecting CYP7A1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18125. Gene ID: 25428. Target Synonyms: CYP7; CYP7-A1; CYPVII; Cholesterol 7 Alpha Hydroxylase; Cytochrome P450 Family 7 Subfamily A Polypeptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2302