Learn More
Biomatik Corporation Human Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) ELISA Kit, 96 tests
Catalog No. 501502303
Encompass_Preferred
The Human Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) ELISA Kit allows for quantitative determination of Cytokeratin Fragment Antigen 21-1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08727. Cross Activity: No significant cross-reactivity or interference between Cytokeratin Fragment Antigen 21-1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2303 Supplier Biomatik Corporation Supplier No. EKU03616
The Human Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) ELISA Kit allows for quantitative determination of Cytokeratin Fragment Antigen 21-1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08727. Cross Activity: No significant cross-reactivity or interference between Cytokeratin Fragment Antigen 21-1 and analogues was observed. Restriction: For Research Use Only.