Learn More
Biomatik Corporation Human D-Amino Acid Oxidase (DAO) ELISA Kit, 96 tests
Catalog No. 501502305
Encompass_Preferred
The Human D-Amino Acid Oxidase (DAO) ELISA Kit allows for quantitative determination of D-Amino Acid Oxidase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14920. Cross Activity: No significant cross-reactivity or interference between D-Amino Acid Oxidase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2305 Supplier Biomatik Corporation Supplier No. EKU03625
The Human D-Amino Acid Oxidase (DAO) ELISA Kit allows for quantitative determination of D-Amino Acid Oxidase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14920. Cross Activity: No significant cross-reactivity or interference between D-Amino Acid Oxidase and analogues was observed. Restriction: For Research Use Only.