Learn More
Biomatik Corporation Human D-Dimer (D2D) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03630
Encompass_Preferred
The Human D-Dimer (D2D) ELISA Kit allows for quantitative determination of D-Dimer in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 61.7-5000 ng/mL. Sensitivity: 26.7 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between D-Dimer and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2306