Learn More
Biomatik Corporation Human D-Dopachrome Tautomerase (DDT) ELISA Kit, 96 tests
Catalog No. 501502309
Encompass_Preferred
The Human D-Dopachrome Tautomerase (DDT) ELISA Kit allows for quantitative determination of D-Dopachrome Tautomerase in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P30046. Cross Activity: No significant cross-reactivity or interference between D-Dopachrome Tautomerase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2309 Supplier Biomatik Corporation Supplier No. EKU03635
The Human D-Dopachrome Tautomerase (DDT) ELISA Kit allows for quantitative determination of D-Dopachrome Tautomerase in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P30046. Cross Activity: No significant cross-reactivity or interference between D-Dopachrome Tautomerase and analogues was observed. Restriction: For Research Use Only.