Learn More
Biomatik Corporation Human Decay Accelerating Factor (DAF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03643
Encompass_Preferred
The Human Decay Accelerating Factor (DAF) ELISA Kit allows for quantitative determination of Decay Accelerating Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08174. Cross Activity: No significant cross-reactivity or interference between Decay Accelerating Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2312