Learn More
Biomatik Corporation Mouse Defensin Alpha 1, Neutrophil (DEFa1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03648
Encompass_Preferred
Mouse Defensin Alpha 1, Neutrophil (DEFa1) ELISA Kit for detecting DEFa1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11477. Gene ID: 13216. Target Synonyms: DEF1; DEFA2; HNP-1; HP-1; MRS; DEF3; HNP-3; HNP3; HP-3; Defensin,Alpha 1; Myeloid-Related Sequence. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2315